missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanine 98.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 5 g |
| Formula Weight | 538.52 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cyanine |
Sécurité et manipulation
| TSCA | No |