missing translation for 'onlineSavingsMsg'
Learn More

Cyanide Standard, 1000 ppm CN-, Ricca Chemical

Catalog No. p-3251604
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
25434 120 mL Amber Poly Bottle
254316 500 mL Amber Poly Bottle
254332 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25434 Supplier Ricca Chemical Company Supplier No. 25434
Only null left

Chemical Identifiers

CAS 151-50-8 , 1310-73-2 , 7732-18-5
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Packaging Amber Poly Bottle
Concentration or Composition Notes 1000ppm CN
Equivalence 1 mL = 1 mg CN
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Cyanide Standard