missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cy™3B Mono-reactive NHS Ester Fluorescent Dyes

Combines exceptional photostability with bright and intense signals. Cytiva Cy™3B NHS Ester Mono-Reactive CyDye™ provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides in fluorescence polarization applications.

$598.00 - $7539.89

Specifications

Formulation 100% Cy3B (NHS ester)
Format Solid
Product Type Cy3B NHS Ester
Molecular Weight (g/mol) 771
Purity >99%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09928193
View Documents
Cytiva
PA63101
1 mg
Each for $598.00
Only null left
 
09928187
View Documents
Cytiva
PA63106
25 mg
Each for $7,539.89
Only null left
 
Description

Description

CyDye™ fluors are versatile fluorophores designed for a broad range of applications. CyDye™ fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye™ flours enables multiplexing without crosstalk for high accuracy.

Cy™3B NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding
Specifications

Specifications

100% Cy3B (NHS ester)
Cy3B NHS Ester
>99%
Red
559
Solid
771
Soluble in aqueous buffers
570
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'