missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cy™3B Mono-reactive NHS Ester Fluorescent Dyes

Catalog No. 09928193
Change view
Click to view available options
Quantity:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09928193 1 mg
09928187 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09928193 Supplier Cytiva Supplier No. PA63101
Only null left

Combines exceptional photostability with bright and intense signals. Cytiva Cy™3B NHS Ester Mono-Reactive CyDye™ provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides in fluorescence polarization applications.

CyDye™ fluors are versatile fluorophores designed for a broad range of applications. CyDye™ fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye™ flours enables multiplexing without crosstalk for high accuracy.

Cy™3B NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding

Specifications

Formulation 100% Cy3B (NHS ester)
Format Solid
Quantity 1 mg
Product Type Cy3B NHS Ester
Molecular Weight (g/mol) 771
Purity >99%
Solubility Soluble in aqueous buffers
Label or Dye Mono-Reactive NHS ester
Color Red
Emission 570
Excitation 559
Show More Show Less