missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Cuvettes for Ciba Corning 550 Express

Catalog No. 361016814
Change view
Click to view available options
Quantity:
100/Cs.
200/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361016814 200/Cs.
361016813 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361016814

Designed for use with the Bayer 550 Express & Express Plus analyzers. Globe Scientific Cuvettes for Ciba Corning 550 Express are packaged in cartridges for easy loading into the analyzers. Each cartridge contains 13 (5-place) cuvette segments.

  • For use with the Bayer 550 Express & Express Plus analyzers
  • Cuvettes are packaged in cartridges for easy loading into the analyzers
  • Each cartridge contains 13 (5-place) cuvette segments
  • Polymethylmethacrylate (PMMA)
  • Natural color
  • Available with 24 disposable bags (21-200-117)

Specifications

Type Cuvette
For Use With (Equipment) Bayer 550 Express & Express Plus Analyzers
Color Natural
Includes Cuvettes only
Quantity 200/Cs.
Material PMMA