Learn More
Type 9M Semi-Micro Black Cuvette with PTFE Cover (Material: Black UV Quartz) (Lightpath: 10mm)
Numéro de catalogue. 501059840
Encompass
Type 9M Semi-Micro Cuvette with PTFE Cover (Material: Black UV Quartz) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Black quartz is to block out and absorb any stray light Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL
Numéro de catalogue. 501059840
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 9M Semi-Micro Cuvette with PTFE Cover (Material: Black UV Quartz) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Black quartz is to block out and absorb any stray light Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL