Learn More
Type 9FL Semi-Micro Fluorescence Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath 10mm)
Numéro de catalogue. 501059830
Encompass
Type 9FL Semi-Micro Fluorescence Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 340-2,500nm) Four windows and the base optically polished for fluorescence work Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL
Numéro de catalogue. 501059830
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 9FL Semi-Micro Fluorescence Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 340-2,500nm) Four windows and the base optically polished for fluorescence work Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL