Learn More
Type 9BM Semi-Micro Raised-Bottom Black Cuvette with PTFE Cover (Material: Black UV Quartz) (Lightpath: 10x1mm)
Numéro de catalogue. 501059824
Encompass
Type 9BM Semi-Micro Raised-Bottom Black Cuvette with PTFE Cover (Material: Black UV Quartz) (Lightpath: 10x1mm) Cuvette has an open top with a PTFE lid which rests on the top Cuvette has a 9mm raised bottom Material is Black UV Quartz (transmits over 80% between 190-2,500nm) Black quartz is to block out and absorb any stray light Two windows optically polished Lightpath is 10x1mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x1x45mm Capacity- 0.4mL
Numéro de catalogue. 501059824
Type 9BM Semi-Micro Raised-Bottom Black Cuvette with PTFE Cover (Material: Black UV Quartz) (Lightpath: 10x1mm) Cuvette has an open top with a PTFE lid which rests on the top Cuvette has a 9mm raised bottom Material is Black UV Quartz (transmits over 80% between 190-2,500nm) Black quartz is to block out and absorb any stray light Two windows optically polished Lightpath is 10x1mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x1x45mm Capacity- 0.4mL