Type 52 Dual Pathlength Fluorescence Cuvette (Material: UV Quartz) (Lightpath: 10x5mm) Cuvette has an open top Is a dual lightpath cell- you can use this cell as a 10mm lightpath cell then rotate it to 90 to use it as a 5mm cell Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished for fluorescence measurements Lightpath is 10x5mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x5x45mm Capacity- 1.75mL