Learn More
Type 52 Dual Pathlength Fluorescence Cuvette (Material: UV Quartz) (Lightpath: 10X1mm)
Numéro de catalogue. 501060253
Encompass
Type 52 Dual Pathlength Fluorescence Cuvette (Material: UV Quartz) (Lightpath: 10X1mm) Cuvette has an open top Is a dual lightpath cell- you can use this cell as a 10mm lightpath cell then rotate it to 90 to use it as a 1mm cell Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished for fluorescence measurements Lightpath is 10x1mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x1x45mm Capacity- 0.35mL
Numéro de catalogue. 501060253
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 52 Dual Pathlength Fluorescence Cuvette (Material: UV Quartz) (Lightpath: 10X1mm) Cuvette has an open top Is a dual lightpath cell- you can use this cell as a 10mm lightpath cell then rotate it to 90 to use it as a 1mm cell Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished for fluorescence measurements Lightpath is 10x1mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x1x45mm Capacity- 0.35mL