Learn More
Type 34 Large Cylindrical Polarimeter Cell with PTFE Stoppers (Material: UV Quartz) (Lightpath: 50mm)
Numéro de catalogue. 501060133
Encompass
Type 34 Large Cylindrical Polarimeter Cell with PTFE Stoppers (Material: UV Quartz) (Lightpath: 50mm) Cuvette has two airtight PTFE stoppers which plug into the top Cell has an elongated cylindrical body design Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 50mm Outside dimensions 52.5x22x35.5mm Chamber Dimensions- 50x19mm Capacity- 14mL
Numéro de catalogue. 501060133
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 34 Large Cylindrical Polarimeter Cell with PTFE Stoppers (Material: UV Quartz) (Lightpath: 50mm) Cuvette has two airtight PTFE stoppers which plug into the top Cell has an elongated cylindrical body design Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 50mm Outside dimensions 52.5x22x35.5mm Chamber Dimensions- 50x19mm Capacity- 14mL