Learn More
Type 21 Macro Cuvette with PTFE Stopper (Material: IR Quartz) (Lightpath: 10mm)
Numéro de catalogue. 501060001
Encompass
Type 21 Macro Cuvette with PTFE Stopper (Material: IR Quartz) (Lightpath: 10mm) Cuvette has an airtight PTFE stopper which plugs into the top Material is IR Quartz (transmits over 80% between 220-3,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x55mm Inside Dimensions- 10x10x45mm Capacity- 3.5mL
Numéro de catalogue. 501060001
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 21 Macro Cuvette with PTFE Stopper (Material: IR Quartz) (Lightpath: 10mm) Cuvette has an airtight PTFE stopper which plugs into the top Material is IR Quartz (transmits over 80% between 220-3,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x55mm Inside Dimensions- 10x10x45mm Capacity- 3.5mL