Learn More
Type 1 Macro Cuvette with PTFE Cover (Material: Pyrex Glass) (Lightpath: 10mm)
Numéro de catalogue. 501059737
Encompass
Type 1 Macro Cuvette with PTFE Cover (Material: Pyrex Glass) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Pyrex Glass (transmits over 80% between 320-2,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x10x45mm Capacity- 3.5mL
Numéro de catalogue. 501059737
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 1 Macro Cuvette with PTFE Cover (Material: Pyrex Glass) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Pyrex Glass (transmits over 80% between 320-2,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x10x45mm Capacity- 3.5mL