Learn More
Type 1 Macro Cuvette with PTFE Cover (Material: IR Quartz) (Lightpath:1mm)
Numéro de catalogue. 501059752
Encompass
Type 1 Macro Cuvette with PTFE Cover (Material: IR Quartz) (Lightpath:1mm) Cuvette has an open top with a PTFE lid which rests on the top Material is IR Quartz (transmits over 80% between 220-3,500nm) Two windows optically polished Lightpath is 1mm Outside dimensions 3.5x12.5x45mm Inside Dimensions- 1x10x45mm Capacity- 0.4mL
Numéro de catalogue. 501059752
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Type 1 Macro Cuvette with PTFE Cover (Material: IR Quartz) (Lightpath:1mm) Cuvette has an open top with a PTFE lid which rests on the top Material is IR Quartz (transmits over 80% between 220-3,500nm) Two windows optically polished Lightpath is 1mm Outside dimensions 3.5x12.5x45mm Inside Dimensions- 1x10x45mm Capacity- 0.4mL