missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupriéthylénèdiamine, certifié, 1,00 m ±0,02 m, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Cupriethylenediamine |
| Couleur | Bleu |
| CAS % maximal | 77.5, 22.5 |
| Forme physique | Liquide |
| Quantité | 4 L |
| Formule linéaire | C4H16CuN4 |
| Numéro UN | UN1761 |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Poids de formule | 183.74 |
| Niveau | Certifié |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Store in a well-ventilated place. Keep container tightly closed. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Réfrigérée |