missing translation for 'onlineSavingsMsg'
Learn More

Cupric Oxide, Black, Powder, CP, 96%, Spectrum™ Chemical

$325.47 - $11023.03

Chemical Identifiers

CAS 1317-38-0
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610016
View Documents
Spectrum Chemical Mfg Cor
C1417125GM
125 g Amber Glass Bottle
Each for $325.47
Only null left
 
18610019
View Documents
Spectrum Chemical Mfg Cor
C1417500GM
500 g Amber Glass Bottle
Each for $557.21
Only null left
 
18610015
View Documents
Spectrum Chemical Mfg Cor
C141725KG
2.5 kg Amber Glass Bottle
Each for $1,985.29
Only null left
 
18610017
View Documents
Spectrum Chemical Mfg Cor
C141712KG
12 kg Poly Pail
Each for $5,788.06
Only null left
 
18610018
View Documents
Spectrum Chemical Mfg Cor
C141745KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1317-38-0

Specifications

1317-38-0
125 g
Chemically Pure
1
96%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.