missing translation for 'onlineSavingsMsg'
Learn More

Cupric Oxide, Black, Powder, CP, 96%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C141725KG

 View more versions of this product

Catalog No. 18610015


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1317-38-0
2.5 kg
Chemically Pure
1
96%
Amber Glass Bottle

Chemical Identifiers

1317-38-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.