missing translation for 'onlineSavingsMsg'
Learn More

Cupric Oxide, Black, Powder, CP, 96%, Spectrum™ Chemical

Catalog No. 18610016
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610016 125 g Amber Glass Bottle
18610019 500 g Amber Glass Bottle
18610015 2.5 kg Amber Glass Bottle
18610017 12 kg Poly Pail
18610018 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610016 Supplier Spectrum Chemical Mfg Cor Supplier No. C1417125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 1317-38-0

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 96%
Grade Chemically Pure
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.