missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupri-Citric Solution R, Ricca Chemical
Specifications
Specifications
| Color | Blue |
| Quantity | 120 mL |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Grade | Laboratory |
| Chemical Name or Material | Cupri-Citric Solution, Reagent |