missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis III Precision Balance, Scale Division 50 mg

Numéro de catalogue. 14561498
Modifier l'affichage
Click to view available options
Capacité d'absorption:
1.5 kg, 3.0 kg, 6.2 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacité d'absorption
14561498 1.5 kg, 3.0 kg, 6.2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 14561498 Fournisseur Sartorius Code fournisseur CUB6202P1S20

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Electronic precision balance with 7 in. color TFT touch display.

  • Deliver precise, reliable weighing results for virtually any regulated lab application, with capacities from 2.1 g to 70 kg and readabilities from 0.1 μg to 1 g.
  • A flexible range of weighing modules, draft shields, and software options lets customers tailor the balances to their workflow, compliance, and integration needs.
  • Upgradeable hardware and software protect your investment and keep your solution ready for future requirements.
  • Built for digital labs, Cubis III enables secure, plug-and-play integration with LIMS, ELN, and other lab systems via standards such as OPC UA, LADS, and SiLA.
  • This helps simplify onboarding of new balances, accelerates validation, and reduces integration costs, while providing a scalable foundation for automation.
  • As connectivity becomes essential to GxP operations, Cubis III delivers real business value by strengthening data integrity, improving audit readiness, accelerating batch release, reducing deviations, and lowering costs.
  • Fits seamlessly into connected labs for a more efficient, integrated, future-ready workflow.

Spécifications

Capacité d'absorption 1.5 kg, 3.0 kg, 6.2 kg
Type d’écran Color TFT Touchscreen
Précision de lecture 10 mg, 20 mg, 50 mg
Catégorie Precision Balance