missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Viral Production Medium

Numéro de catalogue. A5416001
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
100 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A5416001 20 L
A5144001 1 L
A5144002 6 x 1 L
A5144003 10 L
A5416002 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. A5416001 Fournisseur Gibco™ Code fournisseur A5416001
Il en reste null

Gibco CTS Viral Production Medium is a chemically defined, serum-free, protein-free, animal-origin-free, and phenol-red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System.

Gibco CTS Viral Production Medium is a chemically defined, serum-free, protein-free, animal-origin-free, and phenol-red-free medium that is part of the CTS AAV-MAX Helper-Free AAV Production System. CTS Viral Production Medium is specifically developed for growth and transfection of CTS Viral Production Cells 2.0 (suspension-adapted HEK293F-derived cells) for adeno-associated virus (AAV) production.

Features of CTS Viral Production Medium include:

  • Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Formulated with GlutaMAX Supplement, so does not require further supplementation. With the exception of containing GlutaMAX Supplement, the formulation of this product is identical to research-grade Viral Production Medium.
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS AAV-MAX Transfection Kit (A5427701), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

Spécifications

Cell Line Viral Production Cells 2.0
Cell Type HEK293
Classification Animal Origin-free
Form Liquid
Product Type AAV Production Medium
Serum Level Serum-free
Sterility Sterile-filtered
With Additives GlutaMAX
For Use With (Application) AAV Production
Quantity 20 L
Shipping Condition Ambient
Format BPC
Content And Storage 20 L bag. Store at 2°C to 8°C and protect from light.
Culture Type Suspension Cell Culture
Expression System Mammalian
Afficher plus de résultats Afficher moins de résultats