missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ Neurobasal™ Medium
GREENER_CHOICE

Numéro de catalogue. a1371201
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A1371201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A1371201 Fournisseur Gibco™ Code fournisseur A1371201
Il en reste null

CTS Neurobasal Medium is a regulatory-compliant medium for neural stem cell and neuron growth, intended for regenerative medicine applications.

CTS Neurobasal Medium is a chemically defined medium specifically designed to support the growth and viability of pure populations of neural stem cells and PSC-derived neurons. Leveraging a classic formulation trusted by neuroscientists for over 30 years, this medium enhances neuronal survival and differentiation. The CTS version adheres to stringent quality standards and includes comprehensive regulatory documentation, making it ideal for cell therapy manufacturing and clinical research applications.

Benefits of CTS Neurobasal Medium include:

  • Enhanced neuronal support—proven to support the survival and differentiation of neurons
  • Serum-free—eliminates the need for serum or an astrocyte feeder layer when used with CTS B-27 Supplement XenoFree
  • High-quality standards—manufactured at FDA-registered sites that conform to cGMP for medical devices (21 CFR Part 820) and are ISO 13485 and ISO 9001 certified
  • Regulatory compliance—comes with specific intended-use statements, full documentation traceability, and easy access to our Drug Master File (DMF)

Why choose Gibco CTS products?

Gibco CTS products are designed to provide you with a proven choice for transitioning your cell therapy to the clinic with confidence. Our products come with the necessary documentation to support your regulatory filings, ensuring a smooth and compliant process.

Spécifications

Cell Type Neural Cells
Classification Animal Origin-free
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 500 mL
Shipping Condition Ambient
Format Bottle
Content And Storage Store at 2°C to 8°C and protect from light.
Afficher plus de résultats Afficher moins de résultats

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.