missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ AAV-MAX Transfection Kit

Catalog No. A5427701
Change view
Click to view available options
Quantity:
1 Kit (For 1 L of production)
1 Kit (For 10 L of AAV production)
1 Kit (For 100 L of production)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5427701 1 Kit (For 1 L of production)
A5427702 1 Kit (For 10 L of AAV production)
A5427703 1 Kit (For 100 L of production)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5427701 Supplier Gibco™ Supplier No. A5427701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Gibco™ CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System.

The Gibco™ CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It is used for the transient transfection of CTS Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from CTS Viral Production Cells 2.0 when cultured in CTS Viral Production Medium.

Features of the CTS AAV-MAX Transfection Kit include:

  • Components work synergistically within the CTS AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
  • Robust transfection protocols adaptable to large scale transfection
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and following USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

Components of the kit include:

  • CTS AAV-MAX Transfection Reagent – cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
  • CTS AAV-MAX Booster – chemically defined component that works with Transfection Reagent to achieve high transfection efficiency
  • CTS AAV-MAX Enhancer – chemically defined component that enhances the production of AAV particles

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

Specifications

Content And Storage • 1 x 6 mL bottle of CTS AAV-MAX Transfection Reagent
• 1 x 3 mL bottle of CTS AAV-MAX Transfection Booster
• 1 x 10 mL bottle of CTS AAV-MAX Enhancer

Store at 2°C to 8°C. Do not freeze.

Format Kit
Transfection Technique Lipid-based Transfection, Transient Transfection
Expression System Mammalian
Packaging Type Bottle
Product Line AAV-MAX
Product Type Transfection kit for AAV production
Phenol Red Indicator No
Quantity 1 Kit (For 1 L of production)