missing translation for 'onlineSavingsMsg'
Learn More

CSN1 Polyclonal Antibody, Bethyl Laboratories

Numéro de catalogue. 501556758
Encompass
Modifier l'affichage
Click to view available options
Quantité:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
501556758 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 501556758 Fournisseur Bethyl Laboratories Inc Code fournisseur A300026AT

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Spécifications

Antigène CSN1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugué Unconjugated
Formule TBS with 0.1% BSA and 0.09% sodium azide
Gène GPS1
Numéro d’acquisition du gène Q13098
Alias du gène COPS1, CSN1, MGC71287
Symboles de gène GPS1
Espèces hôtes Rabbit
Immunogène Between 400 and C-term
Méthode de purification Antigen affinity chromatography
Quantité 20 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 2873
Espèces cibles Human
Contenu et stockage 4° C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats