missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Violet cristallin, 1% (p/v) en acide acétique glaciaire, pour les titrations non aqueuses, certifié, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Crystal Violet |
| Nom Remarque | 1% (m/v) d’acide acétique glaciaire, pour les titrations non aqueuses |
| Point d’ébullition | 118 °C |
| CAS % maximal | 99, 1 |
| Désignation | Test de réussite |
| Odeur | Vinaigre |
| Emballage | Verre ambré |
| Gravité spécifique | 1.06g/mL |
| Aptitude | Test de réussite |
| Formule linéaire | C25H30CIN3 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Causes severe skin burns and eye damage. Suspected of causing cancer. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Keep away from heat, sparks, open flames, hot surfaces. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, ventilating, lighting equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhahled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up in a cool, well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |