missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Cryostore™ T314-225 Series Storage Boxes

Catalog No. 23045231
Change view
Click to view available options
Holds:
25 (1 to 2 mL Cryogenic Tubes)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
23045231 25 (1 to 2 mL Cryogenic Tubes)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23045231 Supplier Simport™ Scientific Supplier No. T314225B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cryostore™ T314-225 Series Storage Boxes for internally and externally threaded vials.

  • Made of polycarbonate
  • Transparent cover with numeric grid
  • Vented for drainage, stackable
  • Resistant to temperatures between +121°C to −196°C, but should only be used in the gas phase of liquid nitrogen
  • Autoclavable

Specifications

Color Blue
Dimensions (L x W x H) 76 x 76 x 52 mm
Holds 25 (1 to 2 mL Cryogenic Tubes)
Quantity 48/Cs.
Material Polycarbonate
Product Type Storage Box