Learn More
Stemcell Technologies Inc CRYOSTOR CSB, 1000 ML
Supplier:  Stemcell Technologies Inc 1000239
Encompass
CryoStor CSB is an intracellular-like base formulation from the CryoStor preservation media series. It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO). It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death.
Numéro de catalogue. NC1861481
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.