Learn More
Thermo Scientific™ CryoBoxes™
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes.
$197.60 - $2,635.72
| Catalog Number | Mfr. No. | Color | Holds | Material | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Holds | Material | Price | Quantity & Availability | |||
12565227
![]() |
Thermo Scientific™
330821 |
White | 64 (1.5 mL Microcentrifuge Tubes) | Polycarbonate |
Pack of 4 Each for $417.21
Case of 6 Pack for $2,503.26
|
|
|||
1256703
![]() |
Thermo Scientific™
340053 |
White | 100 (1 to 1.8 mL Vials) | Cardboard |
Case of 42 Each for $2,635.72
|
|
|||
1256704
![]() |
Thermo Scientific™
340061 |
White | 100 (3.6 mL Vials) | Cardboard |
Case of 30 Each for $2,234.72
|
|
|||
12565234
![]() |
Thermo Scientific™
341483 |
White | 100 (3.6 mL Vials) | Polycarbonate |
Pack of 4 Each for $294.63
Case of 6 Pack for $1,767.78
|
|
|||
1256701
![]() |
Thermo Scientific™
369639 |
White | 81 (1 to 1.8 mL Vials) | Cardboard |
Case of 48 Each for $2,042.60
|
|
|||
1256702
![]() |
Thermo Scientific™
369647 |
White | 81 (3.6 mL Vials) | Cardboard |
Case of 30 Each for $1,372.19
|
|
|||
12565174
![]() |
Thermo Scientific™
374187 |
Gray | 100 (1 to 1.8 mL Vials) | Polycarbonate |
Pack of 4 Each for $415.21
Case of 6 Pack for $2,491.26
|
|
|||
12565182
![]() |
Thermo Scientific™
378220 |
Brown | 81 (1 to 1.8 mL Vials) | Cardboard |
Case of 6 Each for $197.60
|
|
|||
12565183
![]() |
Thermo Scientific™
378247 |
White | 25 (1 to 1.8 mL Vials) | Cardboard |
Case of 24 Each for $631.88
|
|
|||
15350107
![]() |
Thermo Scientific™
50250505 |
White | 25 (1.2 and 2 mL Vials) | Polycarbonate |
Pack of 8 Each for $363.14
Case of 6 Pack for $2,178.84
|
|
|||
15350107B
![]() |
Thermo Scientific™
50260909 |
White | 81 (1.2 and 2 mL Vials) | Polycarbonate |
Pack of 4 Each for $284.25
Case of 6 Pack for $1,705.50
|
|
|||
033377AA
![]() |
Thermo Scientific™
50261010 |
Blue | 100 (1 to 1.5 mL Vials) | Polycarbonate |
Case of 10 Each for $778.00
|
|
|||
033377J
![]() |
Thermo Scientific™
50270909 |
White | 81 (5 mL Vials) | Polycarbonate |
Pack of 4 Each for $415.61
Case of 6 Pack for $2,493.66
|
|
|||
|
1417560
|
Thermo Scientific™
8670130240 |
Red | 81 (1 to 2 mL Vials) | Polycarbonate |
Case of 24 Each for $1,173.88
|
|
|||
|
1417561
|
Thermo Scientific™
8670130241 |
Yellow | 81 (1 to 2 mL Vials) | Polycarbonate |
Case of 24 Each for $1,163.98
|
|
|||
|
1417562
|
Thermo Scientific™
8670130242 |
Green | 81 (1 to 2 mL Vials) | Polycarbonate |
Case of 24 Each for $1,682.57
|
|
|||
|
1417563
|
Thermo Scientific™
8670130243 |
Blue | 81 (1 to 2 mL Vials) | Polycarbonate |
Case of 24 Each for $1,239.63
|
|
|||
|
1417564
|
Thermo Scientific™
8670130244 |
Gray | 81 (1 to 2 mL Vials) | Polycarbonate |
Case of 24 Each for $1,776.80
|
|
|||
1417565
![]() |
Thermo Scientific™
8670130245 |
Assorted | 81 (1 to 2 mL Vials) | Polycarbonate |
Case of 24 Each for $1,135.36
|
|
|||
Description
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimized to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.
- Durable, all-plastic CryoBoxes™ for short- or long-term storage
- Keyed lids on plastic CryoBoxes™ prevent misalignment
- Plastic boxes are autoclavable for decontamination and usable at temperature range from -196°C to +121°C
- Printed grid on plastic lids for easy inventory management
- Economical chipboard/cardboard CryoBoxes™ for short term storage
- Chipboard/cardboard boxes have dividers and vents in base for good drainage and air circulation
- Available with dividers from 25 to 169 tube storage
Specifications
| Square | |
| Cryo Box |