missing translation for 'onlineSavingsMsg'
Learn More

Crotonaldehyde 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™

Catalog No. C13321G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13321G Supplier TCI America Supplier No. C13321G
Only null left

Chemical Identifiers

CAS 1527-96-4
Molecular Formula C10H10N4O4
Molecular Weight (g/mol) 250.21
MDL Number MFCD00191336
InChI Key GFUVNGJSSAEZHW-UHFFFAOYSA-N
Synonym crotonaldehyde 2,4-dinitrophenylhydrazone, 1-2e-but-2-en-1-ylidene-2-2,4-dinitrophenyl hydrazine
PubChem CID 44630208
IUPAC Name 1-(but-2-en-1-ylidene)-2-(2,4-dinitrophenyl)hydrazine
SMILES CC=CC=NNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 188°C
Quantity 1 g
UN Number 1325
Formula Weight 250.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Crotonaldehyde 2,4-Dinitrophenylhydrazone