missing translation for 'onlineSavingsMsg'
Learn More

Croton Oil, TCI America™

Catalog No. C04215ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04215ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C04215ML Supplier TCI America Supplier No. C04215ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8001-28-3
MDL Number MFCD00130880

Specifications

Quantity 5 mL
UN Number 2810
Physical Form Liquid
Chemical Name or Material Croton Oil
RTECSNumber GQ6300000
TSCA Yes