missing translation for 'onlineSavingsMsg'
Learn More

Croconic Acid 98.0+%, TCI America™

Catalog No. C14835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14835G 5 g
C14831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14835G Supplier TCI America Supplier No. C14835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 488-86-8
Molecular Formula C5H2O5
Molecular Weight (g/mol) 142.066
MDL Number MFCD00181389
InChI Key RBSLJAJQOVYTRQ-UHFFFAOYSA-N
Synonym croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione #
PubChem CID 546874
IUPAC Name 4,5-dihydroxycyclopent-4-ene-1,2,3-trione
SMILES C1(=C(C(=O)C(=O)C1=O)O)O

Specifications

Melting Point 114°C
Color Yellow
Quantity 5 g
Formula Weight 142.07
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Croconic Acid