missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Crimp seals

Catalog No. 111011964
Change view
Click to view available options
For Use With (Equipment):
12 x 32 mm Crimp Top Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111011964 12 x 32 mm Crimp Top Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111011964 Supplier MilliporeSigma™ Supelco™ Supplier No. 508179

Closure type crimp seal

  • The PTFE/rubber septa are compatible with most organic solvents until pierced and are not recommended for aromatics, carbon disulfides, chlorinated solvents, hydrocarbons.

Specifications

Diameter (Metric) Inner 11 mm
Color Red
Material PTFE
Septum Rubber
No. per Pack 100
Thickness (Metric) 1.0 mm
For Use With (Equipment) 12 x 32 mm Crimp Top Vials