missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cresol Red, sodium salt, pure, water soluble, indicator
CAS: 62625-29-0 | C21H17NaO5S
Chemical Identifiers
| CAS | 62625-29-0 |
|---|---|
| Molecular Formula | C21H17NaO5S |
| MDL Number | MFCD00001618 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC151390050
|
Thermo Scientific Chemicals
151390050 |
5 g | Glass bottle |
N/A
|
|
||||
|
AC151390250
|
Thermo Scientific Chemicals
151390250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 62625-29-0 | |
| C21H17NaO5S | |
| MFCD00001618 |
Specifications
| 62625-29-0 | |
| MFCD00001618 | |
| Authentic | |
| Pure | |
| Brown | |
| Cresol Red, sodium salt |
| C21H17NaO5S | |
| Solubility in water: Soluble | |
| 404.4 | |
| from pH 7.0 (yellow) to pH 8.8 (purple) | |
| Powder |
Safety and Handling
EINECSNumber : 263-654-8
TSCA : TSCA
RUO – Research Use Only