missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red, sodium salt, pure, water soluble, indicator

Catalog No. AC151390050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151390050 5 g Glass bottle
AC151390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151390050 Supplier Thermo Scientific Chemicals Supplier No. 151390050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62625-29-0 | C21H17NaO5S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 62625-29-0
Molecular Formula C21H17NaO5S
MDL Number MFCD00001618

Specifications

Solubility Information Solubility in water: Soluble
Infrared Spectrum Authentic
Formula Weight 404.4
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 7.0 (yellow) to pH 8.8 (purple)
Color Brown
Quantity 5 g
Physical Form Powder
Chemical Name or Material Cresol Red, sodium salt
EINECSNumber 263-654-8
TSCA TSCA

RUO – Research Use Only