missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol Red Sodium Salt, TCI America™
Chemical Identifiers
| CAS | 62625-29-0 |
|---|---|
| Molecular Formula | C21H18O5S |
| MDL Number | MFCD00001618 |
| Synonym | Sodium Cresol Red |
Chemical Identifiers
| 62625-29-0 | |
| C21H18O5S | |
| MFCD00001618 | |
| Sodium Cresol Red |
Specifications
| 62625-29-0 | |
| MFCD00001618 | |
| 404.41 | |
| Cresol Red Sodium Salt |
| C21H18O5S | |
| Sodium Cresol Red | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes