missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red Sodium Salt, TCI America™

Catalog No. S00461G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00461G 1 g
S004625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00461G Supplier TCI America Supplier No. S00461G

Chemical Identifiers

CAS 62625-29-0
Molecular Formula C21H18O5S
MDL Number MFCD00001618
Synonym Sodium Cresol Red

Specifications

Formula Weight 404.41
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Cresol Red Sodium Salt