missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red sodium salt

Catalog No. AAB2136106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2136106 5 g
AAB2136114 25 g
AAB2136122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2136106 Supplier Thermo Scientific Chemicals Supplier No. B2136106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red) | CAS: 62625-29-0 | C21H17NaO5S

Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.

Solubility
Soluble in water, dilute alcohols, acids, alkalies and ethanol.

Notes
Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 62625-29-0
MDL Number MFCD00001618

Specifications

Melting Point ∼250°C (decomposition)
Molecular Formula C21H17NaO5S
Solubility Information Soluble in water,dilute alcohols,acids,alkalies and ethanol.
Quantity 5 g
Formula Weight 404.42
Chemical Name or Material Cresol Red sodium salt
EINECSNumber 263-654-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only