missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red, 0.04% Aqueous, pH 0.2 to 1.8 Red to Yellow, pH 7.2 to 8.8 Yellow to Purple, Certified, LabChem™

Supplier:  LabChem LC135201

Quantity 500 mL

 View more versions of this product

Catalog No. LC135201

Only null left

Description

Description

Specifications

Chemical Identifiers

62625-29-0, 7732-18-5

Specifications

Cresol Red
62625-29-0 , 7732-18-5
C21H17O5SNa
Passes Test
Certified
1g/mL
1g/mL
500 mL
0.04% Aqueous, pH 0.2 to 1.8 Red to Yellow, pH 7.2 to 8.8 Yellow to Purple
99.96, 0.04
Soluble in water
404.42
Poly Bottle
Sulfur compounds; Carbon monoxide; Carbon dioxide
Red
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions