missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red, 0.04% Aqueous, pH 0.2 to 1.8 Red to Yellow, pH 7.2 to 8.8 Yellow to Purple, Certified, LabChem™

Catalog No. p-5389056
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC135207 125 mL
LC135201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC135207 Supplier LabChem Supplier No. LC135207

Chemical Identifiers

CAS 62625-29-0 , 7732-18-5

Specifications

CAS Max % 99.96, 0.04
Linear Formula C21H17O5SNa
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 404.42
Grade Certified
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Red
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Cresol Red, 0.04% Aqueous, pH 0.2 to 1.8 Red to Yellow, pH 7.2 to 8.8 Yellow to Purple
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature