missing translation for 'onlineSavingsMsg'
Learn More

Cresol, CP, Spectrum™ Chemical

$363.94 - $2,713.50

Chemical Identifiers

CAS 1319-77-3
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609962
View Documents
Spectrum Chemical Mfg Cor
CR112500MLGL
500 mL Steel Can N/A
Only null left
 
18609961
View Documents
Spectrum Chemical Mfg Cor
CR1124LTGL
4 L Steel Can N/A
Only null left
 
18609959
View Documents
Spectrum Chemical Mfg Cor
CR11220LTMT
20 L Steel Can N/A
Only null left
 
18609960
View Documents
Spectrum Chemical Mfg Cor
CR112200LTBL
200 L Steel Can N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1319-77-3

Specifications

1319-77-3
191 to 203°C
500 mL
Chemically Pure
1
Steel Can
1.030 - 1.040
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.