missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol, CP, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 191 to 203°C |
| Packaging | Steel Can |
| Quantity | 500 mL |
| Specific Gravity | 1.030 - 1.040 |
| Grade | Chemically Pure |
| Physical Form | Liquid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.