missing translation for 'onlineSavingsMsg'
Learn More

Cresol, CP, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor CR112200LTBL

 View more versions of this product

Catalog No. 18609960

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1319-77-3
191 to 203°C
200 L
Chemically Pure
1
Steel Can
1.030 - 1.040
Liquid

Chemical Identifiers

1319-77-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.