missing translation for 'onlineSavingsMsg'
Learn More

Creatinine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014849
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014849 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014849 Supplier Merck Emd Millipore Supplier No. PHR14621G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Creatinine is a by-product produced during the metabolism of creatine phosphate in muscle. It is excreted through urine, and serum creatinine level is most common indicator of renal health. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 60-27-5
Flash Point 290.00°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C4H7N3 O
MDL Number MFCD00059730
Melting Point 295°C
Synonym Creatinine; 2-Amino-1-methyl-2-imidazolin-4-one; 2-Imino-1-methylimidazolidin-4-one; 2-Imino-N-methylhydantoin
Type Certified Reference Material
Quantity 1 g
Formula Weight 113.12
Grade Certified Reference Material
Show More Show Less