missing translation for 'onlineSavingsMsg'
Learn More

Creatine Hydrate 98.0+%, TCI America™

Catalog No. C039625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C039625G 25 g
C0396100G 100 g
C0396500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C039625G Supplier TCI America Supplier No. C039625G

Chemical Identifiers

CAS 6020-87-7
Molecular Formula C4H11N3O3
Molecular Weight (g/mol) 149.15
MDL Number MFCD00004282
InChI Key MEJYXFHCRXAUIL-UHFFFAOYSA-N
Synonym creatine monohydrate, creatine hydrate, 2-1-methylguanidino acetic acid hydrate, creatine, monohydrate, creatinemonohydrate, unii-9603ln7r2q, glycine, n-aminoiminomethyl-n-methyl-, monohydrate, n-amidinosarcosine monohydrate, n-aminoiminomethyl-n-methylglycine monohydrate, creapure
PubChem CID 80116
IUPAC Name 2-[carbamimidoyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=N)N.O

Specifications

Melting Point 295°C
Color White
Quantity 25 g
Formula Weight 131.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Creatine Hydrate
RTECSNumber MB7706000
TSCA Yes