missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF30 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. laevis, X. tropicalis and Bovine
Inhibition of the nuclear export of poly(A)-containing mRNAs caused by the influenza A virus NS1 protein requires its effector domain. The NS1 effector domain functionally interacts with the cellular 30 kDa subunit of cleavage and polyadenylation specific factor 4, an essential component of the 3′ end processing machinery of cellular pre-mRNAs. In influenza virus-infected cells, the NS1 protein is physically associated with cleavage and polyadenylation specific factor 4, 30kD subunit. Binding of the NS1 protein to the 30 kDa protein in vitro prevents CPSF binding to the RNA substrate and inhibits 3′ end cleavage and polyadenylation of host pre-mRNAs. Thus the NS1 protein selectively inhibits the nuclear export of cellular, and not viral, mRNAs. Multiple alternatively spliced transcript variants that encode different isoforms have been described for this gene.
Spécifications
Spécifications
| Antigène | CPSF30 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | CPSF4 |
| Numéro d’acquisition du gène | O95639, Q8BQZ5 |
| Alias du gène | CPSF30, NAR, NEB1, CPSF 30 kDa subunit, NS1 effector domain-binding protein 1, cleavage and polyadenylation specificity factor 30 kDa subunit, cleavage and polyadenylation specificity factor subunit 4, cleavage-polyadenylation specificity factor, 30kD, neb-1, no arches homolog, no |
| Symboles de gène | CPSF4 |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |