missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CP 690550 citrate

Catalog No. 455610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
455610 10 mg
455650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 455610 Supplier Tocris Bioscience™ Supplier No. 4556/10

Potent JAK inhibitor

Potent JAK inhibitor (IC50 values are 1, 20, and 112 nM for JAK3, JAK2 and JAK1 respectively). Orally active immunosuppressant; exhibits efficacy in rodent rheumatoid arthritis models.

Chemical Identifiers

CAS 540737-29-9
Molecular Formula C22H28N6O8

Specifications

Quantity 10 mg
Formula Weight 504.49
Purity >99%
Molecular Weight (g/mol) 504.5
Chemical Name or Material CP 690550 citrate
Recommended Storage Store at +4°C