missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX4 Recombinant Rabbit Monoclonal Antibody (23GB775)

Rabbit Recombinant Monoclonal Antibody
$656.94 - $1,779.35
Spécifications
| Antigen | COX4 |
|---|---|
| Clone | 23GB775 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
MA554168
![]() |
Invitrogen™
MA554168 |
100 μL |
chaque for $1,779.35
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
MA554169
|
Invitrogen™
MA554169 |
20 μL |
chaque for $656.94
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
COX4 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. COX4 (COX4I1) is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.Spécifications
| COX4 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| COX4I1 | |
| AL024441; C16orf2; C16orf4; COX; cox iv; COX IV-1; Cox IV-2; Cox4; COX4 neighbor; Cox4-1; COX4-2; Cox4a; COX4AL; Cox4b; COX4I1; cox4i1.S; Cox4i2; COX4L2; COX4NB; coxiv; COXIV-1; CoxIV-2; cytochrome c oxidase polypeptide IV; Cytochrome c oxidase subunit 4 isoform 1, mitochondrial; cytochrome c oxidase subunit 4 isoform 2, mitochondrial; cytochrome c oxidase subunit 4I1; cytochrome c oxidase subunit 4I2; cytochrome c oxidase subunit IV; cytochrome c oxidase subunit IV isoform 1; cytochrome c oxidase subunit IV isoform 1 precursor; cytochrome c oxidase subunit IV isoform 1 S homeolog; cytochrome c oxidase subunit IV isoform 2; cytochrome c oxidase subunit IV isoform 2 (lung); cytochrome c oxidase subunit IV precursor EC 1.9.3.1; cytochrome c oxidase subunit IV-like 2; cytochrome c oxidase, subunit 4a; cytochrome c oxidase, subunit 4b; cytochrome c oxidase, subunit IV; cytochrome c oxidase, subunit IVa; cytochrome c oxidase, subunit IVb; cytochrome c oxydase subunit 4; dJ857M17.2; EMC8; ER membrane protein complex subunit 8; FAM158B; family with sequence similarity 158, member B; FLJ23483; hypothetical protein MGC73355; IV-1; mg:bb02d03; MGC72016; Neighbor of COX4; NOC4; Protein FAM158B; XELAEV_18024950mg; zgc:110058; zgc:73355 | |
| COX4I1 | |
| Primary | |
| Affinity chromatography |
| 23GB775 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13073 | |
| 1327 | |
| A synthetic peptide of human COX IV (15-53AA). | |
| Antibody |