Learn More
Chemglass Life Sciences Cover Slip, Round, 15mm Diameter, #1 Corning 0211 Glass, 1oz/Pkg, Approx 378/Oz
Supplier: Chemglass Inc CLS1763015
Cover Slip, Round, 15mm Diameter, #1 Corning 0211 Glass, 1oz/Pkg, Approx 378/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1 thickness (0.13 to 0.16mm). Sold in packages of 1oz.5mm Diameter will fit in a 96 well plate; CLS-3500-09612mm Diameter will fit in a 24 well plate; CLS-3500-02415mm Diameter will fit in a 12 well plate; CLS-3500-01225mm Diameter will fit in a 6 well plate; CLS-3500-006