Learn More
Electron Microscopy Circular Cover Glass #1.5, 22 mm, 178 Pieces/oz 1 OZ
Supplier: Electron Microscopy Sciences 7222401
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 22 mm
Package: 178 Pieces/oz
Thickness: 0.16-0.19 mm