Learn More
OHAUS Courier 7000 Series Shipping Scale
The Courier 7000 Series is designed specifically for the needs of manufacturing, distribution, and office shipping. The Courier 7000 Series features intuitive integration with PC-based shipping software so that data is captured accurately and efficiently, every time.
Supplier: OHAUS 30745878
Description
- Designed to withstand constant use from offices to distribution centers. Fast setup with connection to shipping systems like UPS and FedEx.
- GeoCode application for local calibration adjustment.
- Dynamic Weighing retains weight data and presents it on a large, easily accessible backlit display.
- Its WeighAlert mode alerts users to packages that may be too heavy in relation to defined values.
- Courier 7000 can run on USB type-C or disposable battery power.
- Optional Ethernet, second RS232, Bluetooth/WiFi, remote display, ball top and roller top for material handling efficiency are available.
Specifications
| 125 kg | |
| 20 g | |
| Scale |
| LCD | |
| i-C71M125L | |
| Class III |